Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, Silicone Rubber Liner, 34mL, 20x150mm, 100/Pk
SDP
Catalog No. 5000662921
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed to provide dependable performance for sample handling, storage, and culturing in microbiology, clinical diagnostics, and general laboratory environments. Constructed from high quality glass, these tubes feature durable phenolic screw caps that deliver a secure, leak resistant seal and withstand repeated sterilization cycles and daily laboratory use. The flat bottom design allows tubes to stand upright on laboratory benches and work surfaces without requiring additional racks, improving stability during filling and labeling procedures. An integrated white marking area provides a convenient surface for clear sample identification and traceability, helping maintain organized laboratory workflows. These tubes are compatible with standard laboratory racks and accessories and are well suited for culturing, reagent storage, and sample preparation. Each tube includes a silicone rubber liner that enhances sealing reliability.
Catalog No. 50-006-62921 Supplier Alkali Scientific Supplier No. STSC34F
CellPro™ Screw Cap Culture Tubes are designed to provide dependable performance for sample handling, storage, and culturing in microbiology, clinical diagnostics, and general laboratory environments. Constructed from high quality glass, these tubes feature durable phenolic screw caps that deliver a secure, leak resistant seal and withstand repeated sterilization cycles and daily laboratory use. The flat bottom design allows tubes to stand upright on laboratory benches and work surfaces without requiring additional racks, improving stability during filling and labeling procedures. An integrated white marking area provides a convenient surface for clear sample identification and traceability, helping maintain organized laboratory workflows. These tubes are compatible with standard laboratory racks and accessories and are well suited for culturing, reagent storage, and sample preparation. Each tube includes a silicone rubber liner that enhances sealing reliability.