Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, Silicone Rubber Liner, 5mL, 12X75mm, 250/Pk
SDP
Catalog No. 5000662725
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed for dependable sample handling, storage, and culturing in microbiology, clinical, and general laboratory environments. Manufactured from high quality glass, these tubes feature durable phenolic screw caps that provide a secure, leak resistant seal and withstand repeated handling and sterilization cycles. The flat bottom design allows tubes to stand upright on benches and work surfaces without additional racks, improving stability during filling, labeling, and storage. An integrated white marking area enables clear sample identification and traceability, supporting organized workflows in busy laboratories. Compatible with standard racks and accessories, these tubes are ideal for routine culturing, reagent storage, and sample preparation. Each tube includes a silicone rubber liner to enhance sealing performance and chemical resistance during laboratory use.
Catalog No. 50-006-62725 Supplier Alkali Scientific Supplier No. STSC05F
CellPro™ Screw Cap Culture Tubes are designed for dependable sample handling, storage, and culturing in microbiology, clinical, and general laboratory environments. Manufactured from high quality glass, these tubes feature durable phenolic screw caps that provide a secure, leak resistant seal and withstand repeated handling and sterilization cycles. The flat bottom design allows tubes to stand upright on benches and work surfaces without additional racks, improving stability during filling, labeling, and storage. An integrated white marking area enables clear sample identification and traceability, supporting organized workflows in busy laboratories. Compatible with standard racks and accessories, these tubes are ideal for routine culturing, reagent storage, and sample preparation. Each tube includes a silicone rubber liner to enhance sealing performance and chemical resistance during laboratory use.