Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, Silicone Rubber Liner, 9mL, 13x100mm, 250/Pk
SDP
Catalog No. 5000662442
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed for reliable sample handling, storage, and culturing in microbiology, clinical, and research laboratories. Manufactured from high quality glass, these tubes feature durable phenolic screw caps that provide a tight, leak resistant seal and maintain integrity during repeated handling and sterilization cycles. The flat bottom design enables tubes to stand upright on laboratory benches and work surfaces without requiring additional racks, improving stability during filling and labeling. An integrated white marking area supports clear sample identification and traceability, helping maintain organized workflows. These glass culture tubes are compatible with standard laboratory racks and accessories and are suitable for routine culturing, reagent storage, and sample preparation applications. Each tube includes a silicone rubber liner that enhances sealing performance and chemical compatibility.
Catalog No. 50-006-62442 Supplier Alkali Scientific Supplier No. STSC09F
CellPro™ Screw Cap Culture Tubes are designed for reliable sample handling, storage, and culturing in microbiology, clinical, and research laboratories. Manufactured from high quality glass, these tubes feature durable phenolic screw caps that provide a tight, leak resistant seal and maintain integrity during repeated handling and sterilization cycles. The flat bottom design enables tubes to stand upright on laboratory benches and work surfaces without requiring additional racks, improving stability during filling and labeling. An integrated white marking area supports clear sample identification and traceability, helping maintain organized workflows. These glass culture tubes are compatible with standard laboratory racks and accessories and are suitable for routine culturing, reagent storage, and sample preparation applications. Each tube includes a silicone rubber liner that enhances sealing performance and chemical compatibility.