Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Round Bottom, PTFE (Teflon) Faced Liner, 11mL, 16x100mm, 100/Pk
SDP
Catalog No. 5000662727
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed to support reliable sample handling, culturing, and storage across microbiology, clinical, and research laboratory environments. Manufactured from high quality glass, these tubes provide excellent clarity, strength, and chemical resistance for dependable laboratory use. Durable phenolic screw caps with PTFE (Teflon) faced liners form a tight, leak resistant seal that helps protect samples and reagents during storage, handling, or transport. Each tube includes a white marking area to allow easy labeling and sample identification, supporting accurate record keeping in busy laboratory settings. The round bottom design encourages efficient mixing and uniform sample contact, making these tubes suitable for microbial culture growth, reagent preparation, and routine laboratory workflows.
Catalog No. 50-006-62727 Supplier Alkali Scientific Supplier No. STSC11PTFE
CellPro™ Screw Cap Culture Tubes are designed to support reliable sample handling, culturing, and storage across microbiology, clinical, and research laboratory environments. Manufactured from high quality glass, these tubes provide excellent clarity, strength, and chemical resistance for dependable laboratory use. Durable phenolic screw caps with PTFE (Teflon) faced liners form a tight, leak resistant seal that helps protect samples and reagents during storage, handling, or transport. Each tube includes a white marking area to allow easy labeling and sample identification, supporting accurate record keeping in busy laboratory settings. The round bottom design encourages efficient mixing and uniform sample contact, making these tubes suitable for microbial culture growth, reagent preparation, and routine laboratory workflows.