Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Round Bottom, Silicone Rubber Liner, 20mL, 16x150mm, 100/Pk
SDP
Catalog No. 5000662700
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are developed for dependable sample handling, culturing, and storage in microbiology, clinical, and research laboratories. Manufactured from high quality glass, these tubes offer excellent clarity, durability, and resistance to common laboratory chemicals. Each tube features a phenolic screw cap with a silicone rubber liner that forms a tight, leak resistant seal to protect samples and reagents during handling, storage, and transport. A white marking area provides space for easy labeling and accurate sample identification, supporting efficient laboratory organization. The round bottom design promotes effective mixing and uniform sample interaction, making these culture tubes suitable for microbial culture growth, reagent preparation, and routine laboratory workflows.
Catalog No. 50-006-62700 Supplier Alkali Scientific Supplier No. STSC20
CellPro™ Screw Cap Culture Tubes are developed for dependable sample handling, culturing, and storage in microbiology, clinical, and research laboratories. Manufactured from high quality glass, these tubes offer excellent clarity, durability, and resistance to common laboratory chemicals. Each tube features a phenolic screw cap with a silicone rubber liner that forms a tight, leak resistant seal to protect samples and reagents during handling, storage, and transport. A white marking area provides space for easy labeling and accurate sample identification, supporting efficient laboratory organization. The round bottom design promotes effective mixing and uniform sample interaction, making these culture tubes suitable for microbial culture growth, reagent preparation, and routine laboratory workflows.