Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Round Bottom, Silicone Rubber Liner, 34mL, 20x150mm, 100/Pk
SDP
Catalog No. 5000662690
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed to provide reliable sample containment, culturing, and storage across microbiology, clinical, and research laboratory settings. Manufactured from high quality glass, these tubes deliver excellent clarity, durability, and chemical resistance for dependable laboratory use. Each tube is fitted with a phenolic screw cap and silicone rubber liner that creates a secure, leak resistant seal to help safeguard samples and reagents during storage or transport. A convenient white marking area supports easy labeling and clear sample identification, helping maintain organized laboratory workflows. The round bottom design encourages efficient mixing and uniform sample contact, making these culture tubes suitable for microbial culture growth, reagent preparation, and routine laboratory applications.
Catalog No. 50-006-62690 Supplier Alkali Scientific Supplier No. STSC34
CellPro™ Screw Cap Culture Tubes are designed to provide reliable sample containment, culturing, and storage across microbiology, clinical, and research laboratory settings. Manufactured from high quality glass, these tubes deliver excellent clarity, durability, and chemical resistance for dependable laboratory use. Each tube is fitted with a phenolic screw cap and silicone rubber liner that creates a secure, leak resistant seal to help safeguard samples and reagents during storage or transport. A convenient white marking area supports easy labeling and clear sample identification, helping maintain organized laboratory workflows. The round bottom design encourages efficient mixing and uniform sample contact, making these culture tubes suitable for microbial culture growth, reagent preparation, and routine laboratory applications.