Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Round Bottom, Silicone Rubber Liner, 9mL, 13x100mm, 250/Pk
SDP
Catalog No. 5000662806
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are engineered to support dependable sample storage, handling, and culturing in microbiology, clinical, and research laboratory environments. Manufactured from high quality glass, these tubes provide strength, chemical resistance, and excellent visibility for sample observation. Each tube features a durable phenolic screw cap with a silicone rubber liner that forms a tight, leak resistant seal to safeguard samples and reagents during storage, handling, or transport. A white marking area allows convenient labeling and clear sample identification, supporting efficient laboratory organization. The round bottom design improves mixing and uniform sample interaction, making these tubes well suited for microbial culture growth, reagent preparation, and routine laboratory workflows.
Catalog No. 50-006-62806 Supplier Alkali Scientific Supplier No. STSC09
CellPro™ Screw Cap Culture Tubes are engineered to support dependable sample storage, handling, and culturing in microbiology, clinical, and research laboratory environments. Manufactured from high quality glass, these tubes provide strength, chemical resistance, and excellent visibility for sample observation. Each tube features a durable phenolic screw cap with a silicone rubber liner that forms a tight, leak resistant seal to safeguard samples and reagents during storage, handling, or transport. A white marking area allows convenient labeling and clear sample identification, supporting efficient laboratory organization. The round bottom design improves mixing and uniform sample interaction, making these tubes well suited for microbial culture growth, reagent preparation, and routine laboratory workflows.