Learn More
Alkali Scientific CellPro™ SD minus URA, 2X, 500mL
SDP
Catalog No. 502389057
Encompass_Preferred
Alkali Scientific™ SD minus URA (Synthetic Defined minus Uracil) represents a superior solution for yeast culture and growth in molecular biology research. This 2X concentrated, sterile filtered formulation offers optimal purity and sterility, ensuring reliable and consistent results in experimental procedures. Ideal for uracil-deficient yeast strain selection and maintenance, Alkali Scientific™ SD minus URA empowers researchers with a high-quality medium for yeast-based studies.
Catalog No. 50-238-9057 Supplier Alkali Scientific Supplier No. DP610
Alkali Scientific™ SD minus URA (Synthetic Defined minus Uracil) represents a superior solution for yeast culture and growth in molecular biology research. This 2X concentrated, sterile filtered formulation offers optimal purity and sterility, ensuring reliable and consistent results in experimental procedures. Ideal for uracil-deficient yeast strain selection and maintenance, Alkali Scientific™ SD minus URA empowers researchers with a high-quality medium for yeast-based studies.