Learn More
Alkali Scientific CellPro™ SDS, 10% (Sodium Dodecyl Sulfate), 1L, 12/cs
SDP
Catalog No. 502389367
Encompass_Preferred
Alkali Scientific™ presents CellPro™ SDS, Sodium Dodecyl Sulfate, a high-quality reagent designed for protein denaturation and DNA/RNA extraction applications. With its exceptional purity and consistency, CellPro™ SDS ensures reliable results in molecular biology experiments, including Western blotting, DNA sequencing, and protein analysis. This versatile detergent effectively solubilizes hydrophobic molecules and disrupts non-covalent bonds, making it an essential tool for various laboratory protocols. Trust Alkali Scientific™ CellPro™ SDS for superior performance and reproducibility in your research endeavors.
Catalog No. 50-238-9367 Supplier Alkali Scientific Supplier No. PB0283CS
Alkali Scientific™ presents CellPro™ SDS, Sodium Dodecyl Sulfate, a high-quality reagent designed for protein denaturation and DNA/RNA extraction applications. With its exceptional purity and consistency, CellPro™ SDS ensures reliable results in molecular biology experiments, including Western blotting, DNA sequencing, and protein analysis. This versatile detergent effectively solubilizes hydrophobic molecules and disrupts non-covalent bonds, making it an essential tool for various laboratory protocols. Trust Alkali Scientific™ CellPro™ SDS for superior performance and reproducibility in your research endeavors.