Learn More
Alkali Scientific CellPro™ SDS, 10% Sodium Dodecyl Sulfate, 1L, 12/case
SDP
Catalog No. 503029258
Encompass_Preferred
CellPro™ SDS (Sodium Dodecyl Sulfate) CellPro Sodium dodecyl sulfate (SDS) is an alkyl sulfate, anionic detergent used frequently in protein electrophoresis and protein solubilization methods. It is a component of many cell lysis buffer formulations, including RIPA buffer. Typical SDS or lauryl sulfate preparations are mixtures of several different chain lengths (C10 to C18).
Catalog No. 50-302-9258 Supplier Alkali Scientific Supplier No. PBD0283CS
CellPro™ SDS (Sodium Dodecyl Sulfate) CellPro Sodium dodecyl sulfate (SDS) is an alkyl sulfate, anionic detergent used frequently in protein electrophoresis and protein solubilization methods. It is a component of many cell lysis buffer formulations, including RIPA buffer. Typical SDS or lauryl sulfate preparations are mixtures of several different chain lengths (C10 to C18).