Learn More
Alkali Scientific CellPro™ Sealing Mats for 384 Round Well Plates, 50/cs, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503028490
Encompass_Preferred
Black alphanumeric coding Forms a tight seal in plate wells Perforated mats reseal themselves RNase, DNase and pyrogen free CellPro™ Sealing Mats are produced from high quality, chemically resistant silicone, and are designed to provide an airtight and liquid-tight seal. The material resists tearing and wear, minimizing the potential for particulate debris. They are compatible with temperatures from -40 to 80°C, can be autoclaved and are also prepped for reuse with a 10% bleach wash with an ethanol rinse. They also feature black alphanumeric printing for easy reference and two types of resealable perforations (""+"" and ""-""), RNase, DNase and pyrogen free.
Catalog No. 50-302-8490 Supplier Alkali Scientific Supplier No. MPDS32040
Black alphanumeric coding Forms a tight seal in plate wells Perforated mats reseal themselves RNase, DNase and pyrogen free CellPro™ Sealing Mats are produced from high quality, chemically resistant silicone, and are designed to provide an airtight and liquid-tight seal. The material resists tearing and wear, minimizing the potential for particulate debris. They are compatible with temperatures from -40 to 80°C, can be autoclaved and are also prepped for reuse with a 10% bleach wash with an ethanol rinse. They also feature black alphanumeric printing for easy reference and two types of resealable perforations (""+"" and ""-""), RNase, DNase and pyrogen free.