Learn More
Alkali Scientific CellPro™ Self-Standing 50mL Centrifuge Tube, Conical, Bagged, Teal Cap, Sterile, 25/pk, 500/cs
SDP
Catalog No. 502335985
Encompass_Preferred
This Self-Standing 50 mL centrifuge tube is made of polypropylene for clarity and centrifugal strength. This product is made to meet the general requirements of the laboratory such as having clear printed graduations with the marking area, one-hand operation design. Sterilized by E-beam. It is also DNase, RNase and pyrogen-free that is able to handle raw material comply with USP VI requirements.
Catalog No. 50-233-5985 Supplier Alkali Scientific Supplier No. CN5605
This Self-Standing 50 mL centrifuge tube is made of polypropylene for clarity and centrifugal strength. This product is made to meet the general requirements of the laboratory such as having clear printed graduations with the marking area, one-hand operation design. Sterilized by E-beam. It is also DNase, RNase and pyrogen-free that is able to handle raw material comply with USP VI requirements.