Learn More
Alkali Scientific CellPro™ Serological Pipets, Individually Wrapped, Sterile, 10mL, Bagged, 400/Case
SDP
Catalog No. 503028357
Encompass_Preferred
CellPro serological pipettes, or volumetric transfer pipettes, are a common laboratory instrument which is vital for all laboratory operations involving transferring volumes of liquid of from 1mL to 50mL. They are made from 100% polystyrene and featuere bi-directional graduations along with a generous amount of negative graduations. Individually wrapped Gamma Sterilized Color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug 100% Polystyrene construction RNase/DNase, Pyrogen and Cytotoxin Free
Catalog No. 50-302-8357 Supplier Alkali Scientific Supplier No. SP210BCS
CellPro serological pipettes, or volumetric transfer pipettes, are a common laboratory instrument which is vital for all laboratory operations involving transferring volumes of liquid of from 1mL to 50mL. They are made from 100% polystyrene and featuere bi-directional graduations along with a generous amount of negative graduations. Individually wrapped Gamma Sterilized Color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug 100% Polystyrene construction RNase/DNase, Pyrogen and Cytotoxin Free