Learn More
Alkali Scientific CellPro™ SOC Outgrowth Medium, 10 x 10mL
SDP
Catalog No. 5000662409
Encompass_Preferred
CellPro™ SOC Outgrowth Media is a premium, ultra rich recovery medium designed to maximize plasmid transformation efficiency in E. coli. Formulated with peptides, yeast extract, magnesium salts, and glucose, it provides an optimal environment for competent cells to repair membranes, recover metabolic activity, and express antibiotic resistance markers after heat shock or electroporation. The balanced nutrient composition accelerates early cell recovery, improves viability, and supports higher colony yields on selective plates. Sterile filtered and ready to use, this medium ensures reliable performance in demanding molecular biology workflows. It supports rapid recovery of chemically or electro competent E. coli, enhances early membrane repair and plasmid expression, and promotes efficient outgrowth during cloning and plasmid transformation experiments.
Catalog No. 50-006-62409 Supplier Alkali Scientific Supplier No. DC0002OG
CellPro™ SOC Outgrowth Media is a premium, ultra rich recovery medium designed to maximize plasmid transformation efficiency in E. coli. Formulated with peptides, yeast extract, magnesium salts, and glucose, it provides an optimal environment for competent cells to repair membranes, recover metabolic activity, and express antibiotic resistance markers after heat shock or electroporation. The balanced nutrient composition accelerates early cell recovery, improves viability, and supports higher colony yields on selective plates. Sterile filtered and ready to use, this medium ensures reliable performance in demanding molecular biology workflows. It supports rapid recovery of chemically or electro competent E. coli, enhances early membrane repair and plasmid expression, and promotes efficient outgrowth during cloning and plasmid transformation experiments.