Learn More
Alkali Scientific CellPro™ Solution Reservoir, 4 mL, unique 12 channel column basins, PS, White, V bottom, Sterile, 1/pk, 100/Case
SDP
Catalog No. 503027390
Encompass_Preferred
Sloped bottoms of the  4 mL, unique 12 channel column basins reservoirs are useful for filling multichannel-pipette during both cell culture and immunoassay experiments. Manufactured from modified polystyrene Notched volume label
Catalog No. 50-302-7390 Supplier Alkali Scientific Supplier No. RS511
Sloped bottoms of the  4 mL, unique 12 channel column basins reservoirs are useful for filling multichannel-pipette during both cell culture and immunoassay experiments. Manufactured from modified polystyrene Notched volume label