Learn More
Alkali Scientific CellPro™ Spectrophotometer Cuvette, PS, 10mm light path, 1000/Pk, 1.5mL semi-micro, round opening
SDP
Catalog No. 503027499
Encompass_Preferred
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. • Fits most spectrophotometers and photometers• Polystyrene for use in visible range (340-800nm)• Protective packaging – dust-proof and non-scratch• Absorption reproducibility error of less than ±1%**Caps are available and can be ordered separately.
Catalog No. 50-302-7499 Supplier Alkali Scientific Supplier No. EP4101
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. • Fits most spectrophotometers and photometers• Polystyrene for use in visible range (340-800nm)• Protective packaging – dust-proof and non-scratch• Absorption reproducibility error of less than ±1%**Caps are available and can be ordered separately.