Learn More
Alkali Scientific CellPro™ Spheroid Dish, PS / PE, 35mm x 10mm, Sterile 15/Cs, .20mL Low Binding Treatment
SDP
Catalog No. 5000662755
Encompass_Preferred
The CellPro™ Spheroid Dish is a specialized dish designed for spheroid formation in cell culture experiments. With a low binding treatment and a 0.20mL capacity, this dish ensures optimal performance in laboratory settings. It is 35mm x 10mm in size and comes in sterile packaging with 15 dishes per case. Ideal for research on cellular interactions and behavior, this product provides reliable and high-quality results for spheroid-related studies.
Catalog No. 50-006-62755 Supplier Alkali Scientific Supplier No. SFD352
The CellPro™ Spheroid Dish is a specialized dish designed for spheroid formation in cell culture experiments. With a low binding treatment and a 0.20mL capacity, this dish ensures optimal performance in laboratory settings. It is 35mm x 10mm in size and comes in sterile packaging with 15 dishes per case. Ideal for research on cellular interactions and behavior, this product provides reliable and high-quality results for spheroid-related studies.