Learn More
Alkali Scientific CellPro™ SSC 20X (2M NaCl + 0.3M Sodium Citrate), 500mL
SDP
Catalog No. 502389127
Encompass_Preferred
CellPro™ SSC Buffer 20X stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this buffer offers researchers a reliable solution for nucleic acid hybridization experiments. With its sterile filtered formulation, CellPro™ SSC Buffer ensures consistent and reproducible results, empowering researchers in their molecular biology endeavors.
Catalog No. 50-238-9127 Supplier Alkali Scientific Supplier No. PB0285
CellPro™ SSC Buffer 20X stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this buffer offers researchers a reliable solution for nucleic acid hybridization experiments. With its sterile filtered formulation, CellPro™ SSC Buffer ensures consistent and reproducible results, empowering researchers in their molecular biology endeavors.