Learn More
Alkali Scientific CellPro™ SSC 20X (3M NaCL, 0.3M Sodium Citrate), 1L
SDP
Catalog No. 502389384
Encompass_Preferred
CellPro™ SSC Buffer 20X stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this buffer offers researchers a reliable solution for nucleic acid hybridization experiments. With its sterile filtered formulation, CellPro™ SSC Buffer ensures consistent and reproducible results, empowering researchers in their molecular biology endeavors.
Catalog No. 50-238-9384 Supplier Alkali Scientific Supplier No. DS0324
CellPro™ SSC Buffer 20X stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this buffer offers researchers a reliable solution for nucleic acid hybridization experiments. With its sterile filtered formulation, CellPro™ SSC Buffer ensures consistent and reproducible results, empowering researchers in their molecular biology endeavors.