Learn More
Alkali Scientific CellPro Sterile Saline 0.9 Sod
SDP
Catalog No. 503028245
Encompass_Preferred
CellProSterile Saline CellProSterile Saline pH 7 4 0 9% Sodium Chloride USP Sterile Grade (500mL and 1L sizes only) Sterile Saline is a sterile solution of sodium chloride in water for RUO (Research Use Only) It contains no antimicrobial agents and common in Life Science and Research assays Sodium Chloride 0 9% Standard Normal Saline Sterile and endotoxin tested Ideal for laboratory testing biopharmaceutical R and D Ideal in research testing cell culture and molecular protocols Applicable for 0 9% Sodium Chloride USP Sterile Grade products in 500mL and 1L sizes only Also known as inNormal Salinein inBlood Bank Salinein or inIsotonic Salinein Similar osmolality values as found in blood COVID testing and collection Sterile and endotoxin tested CellProNaCl solutions available in a variety of different formats offer a convenient basis for many upstream and downstream bioprocessing applications Manufactured in ISO 9001 and ISO 13485 certified facility Lot-specific certificate of
Catalog No. 50-302-8245 Supplier Alkali Scientific Supplier No. PB0287CS
CellProSterile Saline CellProSterile Saline pH 7 4 0 9% Sodium Chloride USP Sterile Grade (500mL and 1L sizes only) Sterile Saline is a sterile solution of sodium chloride in water for RUO (Research Use Only) It contains no antimicrobial agents and common in Life Science and Research assays Sodium Chloride 0 9% Standard Normal Saline Sterile and endotoxin tested Ideal for laboratory testing biopharmaceutical R and D Ideal in research testing cell culture and molecular protocols Applicable for 0 9% Sodium Chloride USP Sterile Grade products in 500mL and 1L sizes only Also known as inNormal Salinein inBlood Bank Salinein or inIsotonic Salinein Similar osmolality values as found in blood COVID testing and collection Sterile and endotoxin tested CellProNaCl solutions available in a variety of different formats offer a convenient basis for many upstream and downstream bioprocessing applications Manufactured in ISO 9001 and ISO 13485 certified facility Lot-specific certificate of