Learn More
Alkali Scientific CellPro™ Super Broth + Phosphates 500mL
SDP
Catalog No. 502389222
Encompass_Preferred
Alkali Scientific™ CellPro™ Super Broth + Phosphates is a specialized growth medium optimized for the high-yield production of proteins and plasmids in Escherichia coli cultures. With its enriched formulation containing tryptone, yeast extract, and potassium phosphate, this broth supports enhanced bacterial growth and recombinant protein expression. Through meticulous sterile filtration, CellPro™ Super Broth + Phosphates maintains purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.
Catalog No. 50-238-9222 Supplier Alkali Scientific Supplier No. DC0023
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ Super Broth + Phosphates is a specialized growth medium optimized for the high-yield production of proteins and plasmids in Escherichia coli cultures. With its enriched formulation containing tryptone, yeast extract, and potassium phosphate, this broth supports enhanced bacterial growth and recombinant protein expression. Through meticulous sterile filtration, CellPro™ Super Broth + Phosphates maintains purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.