Learn More
Alkali Scientific CellPro™ SuperBroth + Phosphates + 1.5% agar 500mL in 1L Bottle
SDP
Catalog No. 502389326
Encompass_Preferred
Alkali Scientific™ CellPro™ Super Broth is the best commonly used E. coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.
Catalog No. 50-238-9326 Supplier Alkali Scientific Supplier No. DC0038
Alkali Scientific™ CellPro™ Super Broth is the best commonly used E. coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.