Learn More
Alkali Scientific CellPro™ SuperBroth + Phosphates 1L
SDP
Catalog No. 502389285
Encompass_Preferred
Alkali Scientific™ CellPro™ Super Broth is the best commonly used E. coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.
Catalog No. 50-238-9285 Supplier Alkali Scientific Supplier No. DC0037
Alkali Scientific™ CellPro™ Super Broth is the best commonly used E. coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.