Learn More
Alkali Scientific CellPro™ Syringe Filter, PVDF 25mm, 0.45µm, Sterile, Individually Wrapped, 50/cs
SDP
Supplier:  Alkali Scientific TV46S
Encompass_Preferred
These Syringe Filters consists of a plastic housing with a membrane which serves as a filter for fluids. The filter is designed to press fluid through the syringe filter. Specifications: Ultrasonic welded, 25mm filter diameter for volumes between 10-100mL, filter inlet: luer lock female, filter outlet: luer slip male. Maximum operating pressure for 25mm is 7Bar, recommended to use syringe sizes of 3mL or larger. Pore Size of 0.45 µm optimal for thorough cleaning. PVDF, PolyvinyLideneflouride Characteristics: Hydrophilic membrane with good solvent resistance. Low UV absorbing extractables and low non-specific binding. Applications: PVDF is used for general biological filtration, and filtration of samples where high protein recovery is desired. It has slight hydrophilic properties and can be used for clear filtration and sterilization of aqueous solutions.
Catalog No. 50-233-6238