Learn More
Alkali Scientific CellPro™ TAE, 10X (Tris acetate EDTA), 4L Cubitainer
SDP
Catalog No. 502389232
Encompass_Preferred
Alkali Scientific™ CellPro™ TAE Tris Acetate EDTA is a fundamental reagent in nucleic acid electrophoresis, renowned for its exceptional buffering capacity and DNA solubilization properties. Designed to provide optimal performance and reproducibility, this solution enables precise separation and analysis of DNA fragments in agarose gel electrophoresis. Each batch undergoes rigorous sterile filtration to ensure purity and consistency, empowering researchers with confidence in their molecular biology experiments.
Catalog No. 50-238-9232 Supplier Alkali Scientific Supplier No. DB03314L
Alkali Scientific™ CellPro™ TAE Tris Acetate EDTA is a fundamental reagent in nucleic acid electrophoresis, renowned for its exceptional buffering capacity and DNA solubilization properties. Designed to provide optimal performance and reproducibility, this solution enables precise separation and analysis of DNA fragments in agarose gel electrophoresis. Each batch undergoes rigorous sterile filtration to ensure purity and consistency, empowering researchers with confidence in their molecular biology experiments.