Learn More
Alkali Scientific CellPro™ TBE 20X , 20L
SDP
Supplier:  Alkali Scientific DB221620L
Encompass_Preferred
Alkali Scientific™ CellPro™ TBE (Tris-borate EDTA) solution offers a sterile and filtered formulation of this essential buffer for molecular biology and electrophoresis applications. With its meticulous preparation, this solution ensures purity and consistency, meeting the rigorous demands of scientific research and experimentation. The sterile nature of the solution minimizes the risk of contamination, making it ideal for various molecular biology techniques, including DNA and RNA agarose gel electrophoresis. Additionally, the filtered formulation guarantees clarity and reliability in experimental results, providing researchers with confidence in their findings. Whether used for nucleic acid separation, DNA fragment analysis, or other molecular biology procedures, CellPro™ TBE solution delivers consistent performance and reproducible results, supporting the success of laboratory workflows.
Catalog No. 50-238-8793