Learn More
Alkali Scientific CellPro™ Terrific Agar 100mm
SDP
Catalog No. 502389415
Encompass_Preferred
CellPro™ Terrific Agar is a specialized culture medium used for the propagation of recombinant bacterial strains, particularly Escherichia coli (E. coli), in molecular biology laboratories. This agar medium is a modified version of Terrific Broth (TB) agar, supplemented with glycerol, yeast extract, tryptone, and phosphate buffer. Terrific Agar provides optimal conditions for high-density bacterial growth and protein expression, making it ideal for protein production and purification studies. The 100mm plates are convenient for routine bacterial culture experiments and can support the growth of transformed bacterial cells carrying recombinant DNA constructs.
Catalog No. 50-238-9415 Supplier Alkali Scientific Supplier No. DC0135
CellPro™ Terrific Agar is a specialized culture medium used for the propagation of recombinant bacterial strains, particularly Escherichia coli (E. coli), in molecular biology laboratories. This agar medium is a modified version of Terrific Broth (TB) agar, supplemented with glycerol, yeast extract, tryptone, and phosphate buffer. Terrific Agar provides optimal conditions for high-density bacterial growth and protein expression, making it ideal for protein production and purification studies. The 100mm plates are convenient for routine bacterial culture experiments and can support the growth of transformed bacterial cells carrying recombinant DNA constructs.