Learn More
Alkali Scientific CellPro™ Thioglycollate 500mL Glass
SDP
Catalog No. 502388851
Encompass_Preferred
Alkali Scientific™ CellPro™ Thioglycolate is a specialized medium designed for the cultivation of anaerobic and microaerophilic bacteria. With its unique formulation containing thioglycolate, dextrose, and other nutrients, this medium promotes bacterial growth under low-oxygen conditions. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for microbiological studies and microbial diversity analysis.
Catalog No. 50-238-8851 Supplier Alkali Scientific Supplier No. DC0025
Alkali Scientific™ CellPro™ Thioglycolate is a specialized medium designed for the cultivation of anaerobic and microaerophilic bacteria. With its unique formulation containing thioglycolate, dextrose, and other nutrients, this medium promotes bacterial growth under low-oxygen conditions. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for microbiological studies and microbial diversity analysis.