Learn More
Alkali Scientific CellPro™ Tip Comb, 24, V-Bottom, PP, for KingFisher™ Extraction Platform, 10 bags of 5 pieces, 50/cs, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503027137
Encompass_Preferred
Compatible with KingFisher™ Flex Low-binding, medical-grade polypropylene RNase, DNase free CellPro™ Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc. 
Catalog No. 50-302-7137 Supplier Alkali Scientific Supplier No. TL453
Compatible with KingFisher™ Flex Low-binding, medical-grade polypropylene RNase, DNase free CellPro™ Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc. 