Learn More
Alkali Scientific CellPro™ Tip Comb, 8, V-Bottom, PP, for KingFisher™, 240 bags of 2 pieces, 480 / CS, STERILE
SDP
Catalog No. 503026851
Encompass_Preferred
Compatible with KingFisher™ Flex Low-binding, medical-grade polypropylene RNase, DNase free CellPro™ Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc. 
Catalog No. 50-302-6851 Supplier Alkali Scientific Supplier No. TL452
Compatible with KingFisher™ Flex Low-binding, medical-grade polypropylene RNase, DNase free CellPro™ Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc. 