Learn More
Alkali Scientific CellPro™ Tissue Culture Medical Grade Water, 1L , 10/Case
SDP
Catalog No. 503027416
Encompass_Preferred
Cell culture Medical grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Catalog No. 50-302-7416 Supplier Alkali Scientific Supplier No. DTC01CS
Add to Cart
Add to Cart
Cell culture Medical grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).