Learn More
Alkali Scientific CellPro™ Top Agar, 100mL Bottle
SDP
Catalog No. 502389556
Encompass_Preferred
Alkali Scientific™ Top Agar stands as a pinnacle of quality and reliability in molecular biology research. Crafted to meticulous standards and sterile filtered for purity, this 100mL bottle offers an optimal medium for bacterial culture applications. Ideal for phage plaque assays, bacterial transformation, and other molecular biology techniques, Alkali Scientific™ Top Agar ensures consistent and reproducible results in your laboratory experiments.
Catalog No. 50-238-9556 Supplier Alkali Scientific Supplier No. DP605
Alkali Scientific™ Top Agar stands as a pinnacle of quality and reliability in molecular biology research. Crafted to meticulous standards and sterile filtered for purity, this 100mL bottle offers an optimal medium for bacterial culture applications. Ideal for phage plaque assays, bacterial transformation, and other molecular biology techniques, Alkali Scientific™ Top Agar ensures consistent and reproducible results in your laboratory experiments.