Learn More
Alkali Scientific CellPro™ Transfer or Electro Blotting buffer, 10X, pH 8.5
SDP
Catalog No. 502389434
Encompass_Preferred
Alkali Scientific™ CellPro™ Transfer or Electro Blotting Buffer, 10X, pH 8.5, provides a sterile and concentrated solution optimized for Western blotting and protein transfer applications. With a pH of 8.5, this buffer facilitates efficient protein transfer from polyacrylamide gels to membranes, ensuring accurate and reproducible results. The 10X concentration allows for convenient dilution to achieve the desired working concentration, saving time and effort in the laboratory. Sterilized to maintain purity and eliminate the risk of contamination, CellPro™ Transfer or Electro Blotting Buffer is ready for immediate use in your protein research experiments. Trust in the reliability and consistency of CellPro™ buffers for your molecular biology applications, ensuring high-quality results with every experiment.
Catalog No. 50-238-9434 Supplier Alkali Scientific Supplier No. DB0242
Alkali Scientific™ CellPro™ Transfer or Electro Blotting Buffer, 10X, pH 8.5, provides a sterile and concentrated solution optimized for Western blotting and protein transfer applications. With a pH of 8.5, this buffer facilitates efficient protein transfer from polyacrylamide gels to membranes, ensuring accurate and reproducible results. The 10X concentration allows for convenient dilution to achieve the desired working concentration, saving time and effort in the laboratory. Sterilized to maintain purity and eliminate the risk of contamination, CellPro™ Transfer or Electro Blotting Buffer is ready for immediate use in your protein research experiments. Trust in the reliability and consistency of CellPro™ buffers for your molecular biology applications, ensuring high-quality results with every experiment.