Learn More
Alkali Scientific CellPro™ Tris Glycine SDS Buffer, 10X, 4L Cubitainer
SDP
Catalog No. 502389020
Encompass_Preferred
Alkali Scientific™ CellPro™ Tris Glycine offers a sterile solution ideal for a variety of biochemical and molecular biology applications. Composed of Tris (Tris(hydroxymethyl)aminomethane) and Glycine, this buffer provides optimal conditions for protein electrophoresis, particularly in SDS-PAGE (Sodium Dodecyl Sulfate Polyacrylamide Gel Electrophoresis) systems. The sterile formulation ensures the purity and integrity of your samples, minimizing the risk of contamination and ensuring reliable results. CellPro™ Tris Glycine buffer is versatile and suitable for use in protein separation, transfer, and analysis techniques, making it an essential component in the laboratory for protein research and characterization. Trust in the quality and consistency of CellPro™ Tris Glycine for your protein electrophoresis experiments.
Catalog No. 50-238-9020 Supplier Alkali Scientific Supplier No. DB03484L
Alkali Scientific™ CellPro™ Tris Glycine offers a sterile solution ideal for a variety of biochemical and molecular biology applications. Composed of Tris (Tris(hydroxymethyl)aminomethane) and Glycine, this buffer provides optimal conditions for protein electrophoresis, particularly in SDS-PAGE (Sodium Dodecyl Sulfate Polyacrylamide Gel Electrophoresis) systems. The sterile formulation ensures the purity and integrity of your samples, minimizing the risk of contamination and ensuring reliable results. CellPro™ Tris Glycine buffer is versatile and suitable for use in protein separation, transfer, and analysis techniques, making it an essential component in the laboratory for protein research and characterization. Trust in the quality and consistency of CellPro™ Tris Glycine for your protein electrophoresis experiments.