Learn More
Alkali Scientific CellPro™ Trypsin 0.05% protease solution with porcine trypsin, HBSS, EDTA; without calcium, magnesium, 100ml
SDP
Catalog No. 503029156
Encompass_Preferred
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.
Catalog No. 50-302-9156 Supplier Alkali Scientific Supplier No. FB82
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.