Learn More
Alkali Scientific CellPro™ Trypsin 0.25% protease solution with porcine trypsin, HBSS, EDTA; without calcium, magnesium, 100ml, 10/Case
SDP
Catalog No. 503028903
Encompass_Preferred
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.
Catalog No. 50-302-8903 Supplier Alkali Scientific Supplier No. FB81CS
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.