Learn More
Alkali Scientific CellPro™ Trypsin 0.25% protease solution with porcine trypsin, HBSS, EDTA; without calcium, magnesium, 100ml
SDP
Catalog No. 503027726
Encompass_Preferred
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.
Catalog No. 50-302-7726 Supplier Alkali Scientific Supplier No. FB81
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.