Learn More
Alkali Scientific CellPro™ Trypsin 2.5% (10X) protease solution with porcine trypsin, HBSS; without calcium, magnesium, EDTA, or phenol red, 100ml
SDP
Supplier:  Alkali Scientific FB80
Encompass_Preferred
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.   Ships on blue ice.
Catalog No. 50-302-9161