Learn More
Alkali Scientific CellPro™ Trypsin 2.5% (10X) protease solution with porcine trypsin, HBSS; without calcium, magnesium, EDTA, or phenol red, 100ml, 10/Case
SDP
Catalog No. 503026893
Encompass_Preferred
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.   Ships on blue ice.
Catalog No. 50-302-6893 Supplier Alkali Scientific Supplier No. FB80CS
Trypsin cell detachment solution. Derived from porcine pancreas Gamma irradiated prior to hydration and filling Formulated without calcium and magnesium Portfolio includes various concentrations of Trypsin Protease.   Ships on blue ice.