Learn More
Alkali Scientific CellPro™ Trypsin-EDTA Solution 0.25% in HBSS, w/o Ca/Mg, 100 mL, 10mL, 10/Cs
SDP
Catalog No. 503026901
Encompass_Preferred
This perfect blend of trypsin (0.25%) and EDTA (0.53mM) is combined in calcium-free and magnesium-free Hanks Balanced Salt Solution along with phenol red. It has been shown to be a safe and effective means of dissociating a wide variety of cells in culture. Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. Triple 0.1µm sterile-filtered Porcine parvovirus screened Ships on ice. Store frozen at -20 to -10°C
Catalog No. 50-302-6901 Supplier Alkali Scientific Supplier No. FB85CS
This perfect blend of trypsin (0.25%) and EDTA (0.53mM) is combined in calcium-free and magnesium-free Hanks Balanced Salt Solution along with phenol red. It has been shown to be a safe and effective means of dissociating a wide variety of cells in culture. Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. Triple 0.1µm sterile-filtered Porcine parvovirus screened Ships on ice. Store frozen at -20 to -10°C