Learn More
Alkali Scientific CellPro™ Tryptic Soy Broth, 100ml
SDP
Catalog No. 503027554
Encompass_Preferred
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi.
Catalog No. 50-302-7554 Supplier Alkali Scientific Supplier No. TSB01100
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi.