Learn More
Alkali Scientific CellPro™ Tryptic Soy Broth, 100mL, 48/Case
SDP
Catalog No. 5000662463
Encompass_Preferred
CellPro™ Tryptic Soy Broth (TSB), 100mL, is a general purpose liquid culture medium designed to promote the growth and recovery of a wide spectrum of aerobic and facultative anaerobic microorganisms. Formulated with enzymatic digests of casein and soybean meal, this nutrient rich broth supports reliable microbial growth for laboratory testing and research. It is commonly used for sterility testing, microbial enrichment, environmental monitoring, and routine culture maintenance across clinical, pharmaceutical, food safety, and industrial laboratories. The convenient ready to use 100mL format helps streamline microbiological workflows by eliminating preparation steps and ensuring consistent results. Manufactured under strict quality standards, CellPro™ TSB provides dependable performance for routine microbiological analysis and laboratory quality control procedures.
Catalog No. 50-006-62463 Supplier Alkali Scientific Supplier No. TSB01100CS
CellPro™ Tryptic Soy Broth (TSB), 100mL, is a general purpose liquid culture medium designed to promote the growth and recovery of a wide spectrum of aerobic and facultative anaerobic microorganisms. Formulated with enzymatic digests of casein and soybean meal, this nutrient rich broth supports reliable microbial growth for laboratory testing and research. It is commonly used for sterility testing, microbial enrichment, environmental monitoring, and routine culture maintenance across clinical, pharmaceutical, food safety, and industrial laboratories. The convenient ready to use 100mL format helps streamline microbiological workflows by eliminating preparation steps and ensuring consistent results. Manufactured under strict quality standards, CellPro™ TSB provides dependable performance for routine microbiological analysis and laboratory quality control procedures.