Learn More
Alkali Scientific CellPro™ Tryptic Soy Broth, 1L
SDP
Catalog No. 503027555
Encompass_Preferred
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi.
Catalog No. 50-302-7555 Supplier Alkali Scientific Supplier No. TSB011L
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi.