Learn More
Alkali Scientific CellPro™ Tryptic Soy Broth, 225mL, 20/Case
SDP
Catalog No. 5000662780
Encompass_Preferred
CellPro™ Tryptic Soy Broth (TSB), 225mL, is a nutrient rich liquid culture medium formulated to support the growth and recovery of a broad range of aerobic and facultative anaerobic microorganisms. Prepared with enzymatic digests of casein and soybean meal, this versatile medium supplies essential nutrients that promote reliable microbial proliferation. It is widely used in microbiology laboratories for sterility testing, microbial enrichment, environmental monitoring, and routine culture maintenance. The 225mL ready to use format provides a convenient solution for laboratories requiring larger working volumes while maintaining consistent formulation and performance. Manufactured under strict quality standards, CellPro™ Tryptic Soy Broth delivers dependable results for microbiological quality control, research studies, and industrial testing applications.
Catalog No. 50-006-62780 Supplier Alkali Scientific Supplier No. TSB0122
CellPro™ Tryptic Soy Broth (TSB), 225mL, is a nutrient rich liquid culture medium formulated to support the growth and recovery of a broad range of aerobic and facultative anaerobic microorganisms. Prepared with enzymatic digests of casein and soybean meal, this versatile medium supplies essential nutrients that promote reliable microbial proliferation. It is widely used in microbiology laboratories for sterility testing, microbial enrichment, environmental monitoring, and routine culture maintenance. The 225mL ready to use format provides a convenient solution for laboratories requiring larger working volumes while maintaining consistent formulation and performance. Manufactured under strict quality standards, CellPro™ Tryptic Soy Broth delivers dependable results for microbiological quality control, research studies, and industrial testing applications.