Learn More
Alkali Scientific CellPro™ Tryptics Soy Agar 100mm, 10/Sleeve
SDP
Catalog No. 502389414
Encompass_Preferred
CellPro™ Trypticase Soy Agar is a general-purpose medium used for the cultivation and isolation of a wide range of microorganisms in clinical and laboratory settings. This agar medium is formulated with enzymatic digest of casein (tryptone), enzymatic digest of soybean meal (soytone), sodium chloride, agar, and other nutrients. Trypticase Soy Agar supports the growth of both fastidious and non-fastidious bacteria, making it suitable for various microbiological applications, including environmental monitoring, food testing, and pharmaceutical quality control. The 100mm plates provide a convenient platform for bacterial culture experiments and microbial enumeration.
Catalog No. 50-238-9414 Supplier Alkali Scientific Supplier No. DC0136
CellPro™ Trypticase Soy Agar is a general-purpose medium used for the cultivation and isolation of a wide range of microorganisms in clinical and laboratory settings. This agar medium is formulated with enzymatic digest of casein (tryptone), enzymatic digest of soybean meal (soytone), sodium chloride, agar, and other nutrients. Trypticase Soy Agar supports the growth of both fastidious and non-fastidious bacteria, making it suitable for various microbiological applications, including environmental monitoring, food testing, and pharmaceutical quality control. The 100mm plates provide a convenient platform for bacterial culture experiments and microbial enumeration.