Learn More
Alkali Scientific CellPro™ Tryptone Broth 1L
SDP
Catalog No. 502389336
Encompass_Preferred
Alkali Scientific™ CellPro™ Tryptone Broth is a nutrient-rich growth medium formulated for the cultivation of fastidious microorganisms. With its precise combination of tryptone, yeast extract, and sodium chloride, this broth provides essential nutrients to support microbial growth and metabolism. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, ensuring optimal performance in microbiological studies, antimicrobial susceptibility testing, and bacterial fermentation processes.
Catalog No. 50-238-9336 Supplier Alkali Scientific Supplier No. DC0032
Alkali Scientific™ CellPro™ Tryptone Broth is a nutrient-rich growth medium formulated for the cultivation of fastidious microorganisms. With its precise combination of tryptone, yeast extract, and sodium chloride, this broth provides essential nutrients to support microbial growth and metabolism. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, ensuring optimal performance in microbiological studies, antimicrobial susceptibility testing, and bacterial fermentation processes.