Learn More
Alkali Scientific CellPro™ Vacuum Filtration Flasks PES Membrane, 50mL onto Conical Tube, 0.45um Membrane, Individually Wrapped, Sterile, 12/cs
SDP
Supplier:  Alkali Scientific V05045
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PES 0.45µm.The screw-on threaded funnel has a lid, the filtration section has a graduated nozzle connection that fits different diameter vacuum hoses & the receiving bottle has indentations for easy gripping and a separate sterile wrapped cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy. All components are gamma sterilized and certified RNAse, DNAase and pyrogen free.
Catalog No. 50-233-5937