Learn More
Alkali Scientific CellPro™ Vacuum Filtration Flasks, 250mL, 0.45um Membrane, 12/cs
SDP
Catalog No. 502388733
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PES 0.45µm.The screw-on threaded funnel has a lid, the filtration section has a graduated nozzle connection that fits different diameter vacuum hoses & the receiving bottle has indentations for easy gripping and a separate sterile wrapped cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy. All components are gamma sterilized and certified RNAse, DNAase and pyrogen free.
Catalog No. 50-238-8733 Supplier Alkali Scientific Supplier No. V25045
CellPro™ Filtration Flasks are available in filter medium PES 0.45µm.The screw-on threaded funnel has a lid, the filtration section has a graduated nozzle connection that fits different diameter vacuum hoses & the receiving bottle has indentations for easy gripping and a separate sterile wrapped cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy. All components are gamma sterilized and certified RNAse, DNAase and pyrogen free.