Learn More
Alkali Scientific CellPro™ Vented Cell Culture Treated Flasks, 12.5cm2, 10/Pack, 200/Case
SDP
Catalog No. 503027877
Encompass_Preferred
CellPro Vented Flask Cell Culture promote attachment of cells and allow for expanded growth. The vented caps contain a 0.22µm membrane that allows for exchange of gases while minimizing the risk of contamination. They come in a variety of sizes from 12.5cm2 to 300cm2. Packaged in Zip bags High clarity, 100% virgin polystyrene Frosted writing and clear graduations Stackable Sterilized by E-Beam Vent caps (with breathable 0.22µm membrane) Clear lot number for batch traceability Non-Pyrogenic, DNase/RNase free
Catalog No. 50-302-7877 Supplier Alkali Scientific Supplier No. TVN0012
Add to Cart
Add to Cart
CellPro Vented Flask Cell Culture promote attachment of cells and allow for expanded growth. The vented caps contain a 0.22µm membrane that allows for exchange of gases while minimizing the risk of contamination. They come in a variety of sizes from 12.5cm2 to 300cm2. Packaged in Zip bags High clarity, 100% virgin polystyrene Frosted writing and clear graduations Stackable Sterilized by E-Beam Vent caps (with breathable 0.22µm membrane) Clear lot number for batch traceability Non-Pyrogenic, DNase/RNase free