Learn More
Alkali Scientific CellPro™ Vented Cell Culture Treated Flasks, 300cm2, 18/Case
SDP
Catalog No. 503027874
Encompass_Preferred
CellPro Vented Flask Cell Culture promote attachment of cells and allow for expanded growth. The vented caps contain a 0.22µm membrane that allows for exchange of gases while minimizing the risk of contamination. They come in a variety of sizes from 12.5cm2 to 300cm2. Packaged in Zip bags High clarity, 100% virgin polystyrene Frosted writing and clear graduations Stackable Sterilized by E-Beam Vent caps (with breathable 0.22µm membrane) Clear lot number for batch traceability Non-Pyrogenic, DNase/RNase free
Catalog No. 50-302-7874 Supplier Alkali Scientific Supplier No. TVN0300
CellPro Vented Flask Cell Culture promote attachment of cells and allow for expanded growth. The vented caps contain a 0.22µm membrane that allows for exchange of gases while minimizing the risk of contamination. They come in a variety of sizes from 12.5cm2 to 300cm2. Packaged in Zip bags High clarity, 100% virgin polystyrene Frosted writing and clear graduations Stackable Sterilized by E-Beam Vent caps (with breathable 0.22µm membrane) Clear lot number for batch traceability Non-Pyrogenic, DNase/RNase free